Invitrogen
Manufacturer Code:PA528772
Catalog # PIPA528772
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 310 and 374 of Human P2Y11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: P2Y purinoceptor 11; P2Y11; P2Y11 P2Y purinoceptor 11 purinergic receptor P2Y11; purinergic receptor P2Y, G-protein coupled, 11
Gene Aliases: P2RY11; P2Y11
UniProt ID: (Human) Q96G91
Entrez Gene ID: (Human) 5032
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.