Invitrogen
Manufacturer Code:PA5102684
Catalog # PIPA5102684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human P2RY11 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: P2RY11 P2Y purinoceptor 11 P2Y11 purinergic receptor P2Y G-protein coupled 11 purinergic receptor P2Y11; P2Y purinoceptor 11; P2Y11; purinergic receptor P2Y, G-protein coupled, 11
Gene Aliases: P2RY11; P2Y11
UniProt ID: (Human) Q96G91
Entrez Gene ID: (Human) 5032
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.