Invitrogen
Manufacturer Code:PA534074
Catalog # PIPA534074
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 16 amino acid peptide from 2nd extracellular domain of human P2RY10 / P2Y10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G-protein coupled purinergic receptor P2Y10; P2RY10 p2RY10 p2Y-like receptor Putative P2Y purinoceptor 10 p2Y purinoceptor 10 p2Y10; P2Y-like receptor; P2Y10; purinergic receptor P2Y, G-protein coupled, 10; Putative P2Y purinoceptor 10
Gene Aliases: LYPSR2; P2RY10; P2Y10
UniProt ID: (Human) O00398
Entrez Gene ID: (Human) 27334
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.