R&D Systems
Manufacturer Code:FAB9290R025
Catalog # FAB9290R025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 479509 |
Immunogen | HEK293 human cell line transfected with human P2Y1/P2RY1 Accession # NP_002554 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ADP receptor; ATP receptor; ATP receptor P2 purinoceptor subtype Y1 P2RY1 P2Y purinoceptor 1 P2Y1platelet ADP receptor Purinergic receptor purinergic receptor P2Y G-protein coupled 1; P2 purinoceptor subtype Y1; P2Y purinoceptor 1; P2Y1; platelet ADP receptor; Purinergic receptor; purinergic receptor P2Y, G-protein coupled, 1
Gene Aliases: P2RY1; P2Y1
UniProt ID: (Human) P47900
Entrez Gene ID: (Human) 5028
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.