Invitrogen
Manufacturer Code:PA534072
Catalog # PIPA534072
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 19 amino acid peptide from C-terminus of human P2RY1 / P2Y1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP receptor; ATP receptor; P2 purinoceptor subtype Y1; P2RY1 ATP receptor p2Y1 p2y1r Platelet ADP receptor Purinoceptor p2y1 Purinergic receptor p2 purinoceptor subtype Y1 p2RY1 p2Y purinoceptor 1 p2y-1-r Purinoceptor p2yr1 p2y1 receptor; P2Y purinoceptor 1; P2Y1; platelet ADP receptor; Purinergic receptor; purinergic receptor P2Y, G-protein coupled, 1
Gene Aliases: P2RY1; P2Y1
UniProt ID: (Human) P47900
Entrez Gene ID: (Human) 5028
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.