Novus Biologicals
Manufacturer Code:NBP261748
Catalog # NBP2-61748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7G1D6 |
Immunogen | Purified recombinant fragment of human P2RX7 (AA: 226-452) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: ATP receptor; ATP receptor MGC20089 P2X purinoceptor 7 P2X7P2X7 receptor P2X7R P2Z receptor Purinergic receptor purinergic receptor P2X ligand-gated ion channel 7 purinergic receptor P2X7 variant A; P2X purinoceptor 7; P2X7; P2X7 receptor; P2Z receptor; Purinergic receptor; purinergic receptor P2X, ligand gated ion channel, 7; purinergic receptor P2X, ligand-gated ion channel, 7; purinergic receptor P2X7 variant A
Gene Aliases: P2RX7; P2X7
UniProt ID: (Human) Q99572
Entrez Gene ID: (Human) 5027
Molecular Function:
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.