Invitrogen
Manufacturer Code:PA525581
Catalog # PIPA525581
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 563-592 amino acids from the C-terminal region of human P2RX7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP receptor; P2X purinoceptor 7; P2X7; P2X7 ATP receptor P2X purinoceptor 7 P2X7 receptor P2Z receptor purinergic receptor P2X7 variant A; P2X7 purinoceptor; P2X7 receptor; P2Z receptor; Purinergic receptor; purinergic receptor P2X, ligand gated ion channel, 7; purinergic receptor P2X, ligand-gated ion channel, 7; purinergic receptor P2X7 variant A
Gene Aliases: AI467586; P2RX7; P2X(7); P2X7; P2X7R
UniProt ID: (Human) Q99572, (Mouse) Q9Z1M0
Entrez Gene ID: (Human) 5027, (Mouse) 18439
Molecular Function:
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.