Invitrogen
Manufacturer Code:PA577683
Catalog # PIPA577683
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)HRSHTRLAEMKYE corresponding to amino acid residues 299-311 of rat P2X5 receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Membrane magnesium transporter 2; membrane magnesium transporter 2 {ECO:0000250|UniProtKB:Q8R3L0}; mmgt2; mmgt2 {ECO:0000250|UniProtKB:Q8R3L0}; P2X5 Receptor (extracellular)
Gene Aliases: AI852046; Mmgt2; RGD1311260
UniProt ID: (Rat) Q6P719, (Mouse) Q8R3L0
Entrez Gene ID: (Rat) 303211, (Mouse) 216829
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.