Invitrogen
Manufacturer Code:PA541080
Catalog # PIPA541080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human P2RX4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP receptor; ATP-gated cation channel protein; ionotropic purinergic receptor; P2 purinergic receptor subtype; P2X purinoceptor 4; P2X receptor, subunit 4; P2X4; P2X4 purinoceptor; Purinergic receptor; Purinergic receptor P2X ligand-gated ion channel 4 P2X4 P2X4R; purinergic receptor P2X, ligand gated ion channel, 4; purinergic receptor P2X, ligand-gated ion channel, 4; purinergic receptor P2X4; purinergic receptor P2X4 subunit variant 1; purinergic receptor P2X4 subunit variant 2; purinergic receptor P2x4 subunit variant e; purinoceptor P2X4; putative partial extracellular loop region
Gene Aliases: AI504491; AW555605; D5Ertd444e; P2RX4; P2X4; P2X4R
UniProt ID: (Bovine) Q5E9U1, (Human) Q99571, (Rat) P51577
Entrez Gene ID: (Pig) 100157723, (Rabbit) 100008720, (Bovine) 338036, (Dog) 448783, (Human) 5025, (Guinea pig) 100716323, (Mouse) 18438, (Rat) 29659
Molecular Function: ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.