Invitrogen
Manufacturer Code:PA577682
Catalog # PIPA577682
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KGFGRYANRVMDVSD corresponding to amino acid residues 65-79 of rat P2X3 Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP receptor; P2RX3; P2X purinoceptor 3; P2X3; Purinergic receptor; purinergic receptor P2X3
Gene Aliases: 4930513E20Rik; P2rx3; P2X3
UniProt ID: (Rat) P49654, (Mouse) Q3UR32
Entrez Gene ID: (Rat) 81739, (Mouse) 228139
Molecular Function:
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.