Novus Biologicals
Manufacturer Code:NBP219657
Catalog # NBP219657
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human P2X3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP receptor; ATP receptor MGC129956 P2X purinoceptor 3 P2X receptor subunit 3 P2X3purinergic receptor P2X3 Purinergic receptor purinergic receptor P2X ligand-gated ion channel 3 purinoceptor P2X3; P2X purinoceptor 3; P2X receptor, subunit 3; P2X3; Purinergic receptor; purinergic receptor P2X, ligand gated ion channel, 3; purinergic receptor P2X, ligand-gated ion channel, 3; purinergic receptor P2X3; purinoceptor P2X3
Gene Aliases: P2RX3; P2X3
UniProt ID: (Human) P56373
Entrez Gene ID: (Human) 5024
Molecular Function: ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.