Invitrogen
Manufacturer Code:PA577681
Catalog # PIPA577681
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KSDYLKHSTFDQDS corresponding to amino acid residues 207-220 of rat P2X2 with replacement of cysteine 214 (C214 ) with serine (*S) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP receptor; P2RX2; P2X purinoceptor 2; P2X2; Purinergic receptor
Gene Aliases: P2rx2; P2X2; P2X2a
UniProt ID: (Rat) P49653, (Mouse) Q8K3P1
Entrez Gene ID: (Rat) 114115, (Mouse) 231602
Molecular Function: ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.