Invitrogen
Manufacturer Code:PA528845
Catalog # PIPA528845
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 407 and 471 of Human P2X2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP receptor; P2X purinoceptor 2; P2X Receptor, subunit 2; P2X2; P2X2 ATP receptor P2X Receptor subunit 2 P2X purinoceptor 2; Purinergic receptor; purinergic receptor P2X, ligand gated ion channel, 2; purinergic receptor P2X, ligand-gated ion channel, 2
Gene Aliases: DFNA41; P2RX2; P2X2
UniProt ID: (Human) Q9UBL9
Entrez Gene ID: (Human) 22953
Molecular Function:
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.