Invitrogen
Manufacturer Code:PA124624
Catalog # PIPA124624
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues D(460) S T S T D P K G L A Q L(472) of rat P2X2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP receptor; MGC129601 P2X2; P2X purinoceptor 2; P2X Receptor, subunit 2; P2X2; Purinergic receptor; purinergic receptor P2X, ligand gated ion channel, 2; purinergic receptor P2X, ligand-gated ion channel, 2
Gene Aliases: DFNA41; P2RX2; P2X2; P2X2a
UniProt ID: (Human) Q9UBL9, (Guinea pig) O70397, (Rat) P49653, (Mouse) Q8K3P1
Entrez Gene ID: (Human) 22953, (Pig) 100736581, (Guinea pig) 100135591, (Rat) 114115, (Mouse) 231602
Molecular Function: ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.