Novus Biologicals
Manufacturer Code:NBP286742
Catalog # NB034364
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human P2X1/P2RX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP receptor; ATP receptor P2X purinoceptor 1 P2X receptor subunit 1 P2X1P2X1 receptor Purinergic receptor purinergic receptor P2X ligand-gated ion channel 1; P2X purinoceptor 1; P2X receptor, subunit 1; P2X1; P2X1 receptor; Purinergic receptor; purinergic receptor P2X, ligand gated ion channel, 1; purinergic receptor P2X, ligand-gated ion channel, 1; purinergic receptor P2X1
Gene Aliases: P2RX1; P2X1
UniProt ID: (Human) P51575
Entrez Gene ID: (Human) 5023
Molecular Function: ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.