Invitrogen
Manufacturer Code:PA5103994
Catalog # PIPA5103994
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human P2RX1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI323649 ATP receptor BB122383 P2RX1 P2x P2X purinoceptor 1 P2X receptor subunit 1 P2X1 P2X1 receptor P2XMR Pdcd3 programmed cell death 3 Purinergic receptor purinergic receptor P2X 1 purinergic receptor P2X ligand gated ion channel 1 purinergic receptor P2X ligand-gated ion channel 1 purinergic receptor P2X1 RP-2 RP-2 protein; ATP receptor; P2X purinoceptor 1; P2X receptor, subunit 1; P2X1; P2X1 receptor; programmed cell death 3; Purinergic receptor; purinergic receptor P2X, ligand gated ion channel, 1; purinergic receptor P2X, ligand-gated ion channel, 1; purinergic receptor P2X1; RP-2 protein
Gene Aliases: AI323649; BB122383; P2RX1; P2x; P2X1; P2XMR; Pdcd3; RP-2
UniProt ID: (Human) P51575, (Rat) P47824, (Mouse) P51576
Entrez Gene ID: (Human) 5023, (Rat) 25505, (Mouse) 18436
Molecular Function: ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.