Invitrogen
Manufacturer Code:PA511586
Catalog # PIPA511586
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 114-142 amino acids from the N-terminal region of human CDH3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cadherin 3, type 1, P-cadherin (placental); Cadherin-3; Cadherin-3 Placental cadherin P-cadherin CADH3 CDHP; calcium-dependent adhesion protein, placental; P-cadherin; Placental cadherin; RPE-specific cadherin
Gene Aliases: AI385538; Cadp; CDH3; CDHP; HJMD; P-cadherin; PCAD
UniProt ID: (Human) P22223, (Mouse) P10287
Entrez Gene ID: (Human) 1001, (Mouse) 12560
Molecular Function: cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.