R&D Systems
Manufacturer Code:MAB761SP
Catalog # MAB761SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 106020 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse P-Cadherin Glu100-Gly647 Accession # Q8BSL6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CAD3 cadherin 3 P-cadherin (placental) cadherin 3 type 1 P-cadherin (placental) cadherin-3 CDH3 CDHP CDHPcalcium-dependent adhesion protein placental HJMD PCAD PCadherin PCADP-cadherin Placental cadherin; cadherin 3, type 1, P-cadherin (placental); Cadherin-3; calcium-dependent adhesion protein, placental; P-cadherin; Placental cadherin
Gene Aliases: CDH3; CDHP; HJMD; PCAD
UniProt ID: (Human) P22223
Entrez Gene ID: (Human) 1001
Molecular Function: cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.