Invitrogen
Manufacturer Code:MA537974
Catalog # PIMA537974
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2266 |
Immunogen | A synthesized peptide derived from human Oxytocin Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: OT-R; OTR OT-R OTR1 oxcytocin receptor OXTR Oxt-r Oxytocin receptor; oxcytocin receptor; Oxytocin receptor
Gene Aliases: OT-R; OTR; OTR1; OXTR
UniProt ID: (Human) P30559, (Mouse) P97926, (Rat) P70536
Entrez Gene ID: (Human) 5021, (Mouse) 18430, (Rat) 25342
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.