Invitrogen
Manufacturer Code:PA575431
Catalog # PIPA575431
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 11-60 of Human Oxytocin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neurophysin 1; neurophysin I; Ocytocin; OT-NPI; oxcytocin; Oxytocin; oxytocin, prepro- (neurophysin I); oxytocin, prepropeptide; Oxytocin-neurophysin 1; Oxytocin-neurophysin 1 OT-NPI Oxytocin Ocytocin Neurophysin 1 OXT OT; oxytocin-neurophysin I, preproprotein; oxytocin/neurophysin (Oxt) gene, complete gene, complete cds
Gene Aliases: OT; OT-NPI; OXT; OXT-NPI; Oxy
UniProt ID: (Human) P01178, (Mouse) P35454, (Rat) P01179
Entrez Gene ID: (Human) 5020, (Mouse) 18429, (Rat) 25504
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.