Invitrogen
Manufacturer Code:PA5112637
Catalog # PIPA5112637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Oxytocin-neurophysin 1 protein (27-123AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neurophysin 1; Neurophysin 1 neurophysin I Ocytocin OT OT-NPI oxcytocin Oxt OXT-NPI Oxy Oxytocin oxytocin prepro- (neurophysin I) oxytocin prepropeptide oxytocin/neurophysin (Oxt) gene complete gene complete cds oxytocin/neurophysin 1 prepropeptide oxytocin/neurophysin I prepropeptide Oxytocin-neurophysin 1 oxytocin-neurophysin I oxytocin-neurophysin I preproprotein prepropeptide for oxytocin and neurophysin unnamed protein product; neurophysin I; Ocytocin; OT-NPI; Oxytocin; oxytocin, prepro- (neurophysin I); oxytocin, prepropeptide; Oxytocin-neurophysin 1; oxytocin-neurophysin I, preproprotein
Gene Aliases: OT; OT-NPI; OXT; OXT-NPI; Oxy
UniProt ID: (Human) P01178, (Mouse) P35454
Entrez Gene ID: (Human) 5020, (Mouse) 18429
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.