Novus Biologicals
Manufacturer Code:NBP152151
Catalog # NBP152151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence DGCRTDPACDPES corresponding to C-Terminus (near) according to Uniprot Mouse NP_035155.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: MGC126890 MGC126892 neurophysin I OT OT-NPI oxytocin prepro- (neurophysin I) oxytocin prepropeptide oxytocin-neurophysin 1 oxytocin-neurophysin I preproprotein; Neurophysin 1; neurophysin I; Ocytocin; OT-NPI; Oxytocin; oxytocin, prepro- (neurophysin I); oxytocin, prepropeptide; Oxytocin-neurophysin 1; oxytocin-neurophysin I, preproprotein
Gene Aliases: OT; OT-NPI; OXT; OXT-NPI
UniProt ID: (Human) P01178
Entrez Gene ID: (Human) 5020
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.