Abnova
Manufacturer Code:PAB18646
Catalog # 89-120-204
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids near C-terminus of mouse Oxt . |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of Oxt.
For Research Use Only
Protein Aliases: Neurophysin 1; Ocytocin; OT; OT-NPI; Oxy; Oxytocin; Oxytocin-neurophysin 1
Gene Aliases: OT; Oxt; Oxy
UniProt ID: (Mouse) P35454
Entrez Gene ID: (Mouse) 18429
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.