Novus Biologicals
Manufacturer Code:NB10062809H
Catalog # NB10062809H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Chicken |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C6 |
Immunogen | Spleen cells from Ovalbumin immunized Balb/c mice were fused with cells of the mouse myeloma P3U1 |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Allergen Gal d 2 CAP 2 CAP2 Cytoplasmic antiproteinase 2 Egg albumin Gal d II Hen egg white PI8 Plakalbumin Protease inhibitor 8 Serpin B8 serpin peptidase inhibitor clade B member 8 SERPINB14 SERPINB8; Allergen Gal d II; clade (ovalbumin); Egg albumin; Gal d 2; member 14; Ovalbumin; Plakalbumin; serpin peptidase inhibitor; unnamed protein product
Gene Aliases: OVA; SERPINB14
UniProt ID: (Chicken) P01012
Entrez Gene ID: (Chicken) 396058
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.