Invitrogen
Manufacturer Code:PA1196HRP
Catalog # PIPA1196HRP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Chicken |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Ovalbumin |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Allergen Gal d II; clade (ovalbumin); Egg albumin; Egg albumin SERPINB14 Plakalbumin Allergen Gal d 2 Allergen Gal d II PA1-196HRP; Gal d 2; member 14; Ovalbumin; Plakalbumin; serpin peptidase inhibitor; unnamed protein product
Gene Aliases: OVA; SERPINB14
UniProt ID: (Chicken) P01012
Entrez Gene ID: (Chicken) 396058
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.