Novus Biologicals
Manufacturer Code:NB10056505SS
Catalog # NB10056505T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 98A1071 |
Immunogen | A peptide corresponding to the amino acids 20-37 TQETFPPKYLHYDEETSH of human Osteoprotegerin was used as immunogen for this antibody. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: OCIFMGC29565 OPG OPGtumor necrosis factor receptor superfamily member 11B Osteoclastogenesis inhibitory factor Osteoprotegerin TR1 tumor necrosis factor receptor superfamily member 11b; Osteoclastogenesis inhibitory factor; Osteoprotegerin; Tumor necrosis factor receptor superfamily member 11B; tumor necrosis factor receptor superfamily, member 11b
Gene Aliases: OCIF; OPG; PDB5; TNFRSF11B; TR1
UniProt ID: (Human) O00300
Entrez Gene ID: (Human) 4982
Molecular Function:
cytokine receptor
receptor
tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.