R&D Systems
Manufacturer Code:MAB4591SP
Catalog # MAB4591SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 110511 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Osteoprotegerin/TNFRSF11B Gln21-Leu401 (Gln138Arg) Accession # Q6PI12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: OCIF OCIFMGC29565 OPG OPGtumor necrosis factor receptor superfamily member 11B Osteoclastogenesis inhibitory factor Osteoprotegerin TNFRSF11B TR1 tumor necrosis factor receptor superfamily member 11b; Osteoclastogenesis inhibitory factor; Osteoprotegerin; Tumor necrosis factor receptor superfamily member 11B; tumor necrosis factor receptor superfamily, member 11b
Gene Aliases: OCIF; OPG; PDB5; TNFRSF11B; TR1
UniProt ID: (Human) O00300
Entrez Gene ID: (Human) 4982
Molecular Function: cytokine receptor receptor tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.