Novus Biologicals
Manufacturer Code:NB11089062SS
Catalog # NB11089062T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B20 |
Immunogen | Synthetic peptide corresponding to the C-terminus of human Osteopontin [Swiss-Prot# P10451]. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BNSP Bone sialoprotein 1 MGC110940 Nephropontin osteopontin secreted phosphoprotein 1bone sialoprotein I early T-lymphocyteactivation 1) secreted phosphoprotein-1 (osteopontin bone sialoprotein) SPP-1 SPP1/CALPHA1 fusion Urinary stone protein uropontin; Bone sialoprotein 1; early T-lymphocyte activation 1; Nephropontin; Osteopontin; osteopontin/immunoglobulin alpha 1 heavy chain constant region fusion protein; Secreted phosphoprotein 1; secreted phosphoprotein 1 (osteopontin, bone sialoprotein I, early T-lymphocyte activation 1); secreted phosphoprotein 1 variant 6; SPP-1; SPP1/CALPHA1 fusion; Urinary stone protein; Uropontin
Gene Aliases: BNSP; BSPI; ETA-1; OPN; PSEC0156; SPP1
UniProt ID: (Human) Q567T5
Entrez Gene ID: (Human) 6696
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.