Proteintech
Manufacturer Code:127551AP150UL
Catalog # 50-172-8225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Osteoglycin Fusion Protein Ag3484 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: corneal keratan sulfate proteoglycan; Mimecan; Mimecan OG OGN OIF Osteoglycin Osteoinductive factor SLRR3A; mimecan proteoglycan; OIF; Osteoglycin; Osteoinductive factor
Gene Aliases: 3110079A16Rik; mimecan; mimican; OG; OGN; OIF; SLRR3A
UniProt ID: (Human) P20774, (Mouse) Q62000
Entrez Gene ID: (Human) 4969, (Rat) 291015, (Mouse) 18295
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.