Proteintech
Manufacturer Code:234181AP150UL
Catalog # 50-173-4071
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Osteocalcin Fusion Protein Ag20065 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BGLAP Osteocalcin BGP Bone Gla protein OC OCN Osteocalcin PMF1; BGP; bone gamma carboxyglutamate protein 1; bone gamma-carboxyglutamate (gla) protein (osteocalcin); bone gamma-carboxyglutamate protein 2; bone gamma-carboxyglutamic acid (Gla) protein (osteocalcin); Bone Gla protein; bone Gla-protein; Gamma-carboxyglutamic acid-containing protein; Osteocalcin
Gene Aliases: BGLAP; Bglap1; Bglap2; BGP; Bgpr; Bgpra; mOC-A; OC; OCN; OG1
UniProt ID: (Human) P02818, (Mouse) P86546, (Rat) P04640
Entrez Gene ID: (Human) 632, (Mouse) 12096, (Rat) 25295
Molecular Function:
calcium-binding protein
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.