Proteintech
Manufacturer Code:183701AP150UL
Catalog # 50-173-2133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Orexin receptor 1 Fusion Protein Ag12269 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HCRTR1 hypocretin (orexin) receptor 1 Hypocretin receptor type 1 Orexin receptor 1 Orexin receptor type 1 Ox 1 R Ox1 R OX1R; hypocretin (orexin) receptor 1; hypocretin receptor 1; Hypocretin receptor type 1; hypocretin receptor-1; orexin receptor 1; Orexin receptor type 1; orexin receptor-1; Ox-1-R; ox1-R; ox1r
Gene Aliases: HCRTR1; Hctr1; ox-1-R; ox1-R; OX1R
UniProt ID: (Human) O43613, (Mouse) P58307, (Rat) P56718
Entrez Gene ID: (Human) 3061, (Mouse) 230777, (Rat) 25593
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.