Invitrogen
Manufacturer Code:MA524178
Catalog # PIMA524178
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 456738 |
Immunogen | NS0 mouse myeloma cell line transfected with human Orexin R2/HCRTR2 Met1-Trp444 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HCRTR1 Hcrtr2 OX1R OX2R HCRTR1 hypocretin orexin receptor 1 hypocretin receptor 1 hypocretin receptor-1 orexin receptor 1 orexin receptor-1 HCRTR2 hypocretin orexin receptor 2 hypocretin receptor 2 hypocretin receptor-2 orexin receptor 2 orexin receptor-2; hypocretin (orexin) receptor 2; Hypocretin receptor type 2; Orexin receptor type 2; orexin type-2 receptor; Ox-2-R; ox2-R
Gene Aliases: HCRTR2; OX2R
UniProt ID: (Human) O43614
Entrez Gene ID: (Human) 3062
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.