Invitrogen
Manufacturer Code:PA577567
Catalog # PIPA577567
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)DRLARGRTSTESRKS corresponding to amino acid residues 391-405 of rat OX2 R |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hypocretin (orexin) receptor 2; hypocretin receptor 2; Hypocretin receptor type 2; Orexin receptor type 2; orexin type-2 receptor; Ox-2-R; ox2-R; ox2r; OX2R HCRTR2 Hypocretin Receptor 2
Gene Aliases: HCRTR2; mOX2aR; mOX2bR; Mox2r; mOXR2; OX2R
UniProt ID: (Human) O43614, (Rat) P56719, (Mouse) P58308
Entrez Gene ID: (Human) 3062, (Rat) 25605, (Mouse) 387285
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.