Invitrogen
Manufacturer Code:PA533837
Catalog # PIPA533837
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 14 amino acid peptide from 2nd extracellular domain of human HCRTR1 / Orexin Receptor 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HCRTR1 Hcrt receptor 1 Hypocretin receptor 1 Hypocretin type 1 receptor Hypocretin receptor-1 Orexin receptor type 1 Orexin type 1 receptor Orexin-1 receptor OX1 receptor Orexin a receptor Orexin receptor-1 OX-R1 Orexin receptor 1 Ox1-R OX1R Hypocretin (orexin) receptor 1 Hypocretin receptor type 1 Ox-1-R; hypocretin (orexin) receptor 1; hypocretin receptor 1; Hypocretin receptor type 1; hypocretin receptor-1; orexin receptor 1; Orexin receptor type 1; orexin receptor-1; Ox-1-R; ox1-R; ox1r
Gene Aliases: HCRTR1; Hctr1; OX1R
UniProt ID: (Human) O43613, (Rat) P56718
Entrez Gene ID: (Human) 3061, (Rat) 25593
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.