R&D Systems
Manufacturer Code:MAB52461SP
Catalog # MAB52461SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 456723 |
Immunogen | NS0 mouse myeloma cell line transfected with human Orexin R2/HCRTR2 Met1-Trp444 Accession # AAC39602 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HCRTR2 hypocretin (orexin) receptor 2 Hypocretin receptor type 2 Orexin Receptor 2 orexin receptor type 2 OrexinR2 Ox-2-R Ox2-R OX2R; hypocretin (orexin) receptor 2; Hypocretin receptor type 2; Orexin receptor type 2; orexin type-2 receptor; Ox-2-R; ox2-R
Gene Aliases: HCRTR2; OX2R
UniProt ID: (Human) O43614
Entrez Gene ID: (Human) 3062
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.