Novus Biologicals
Manufacturer Code:NBP295106100UL
Catalog # NB024374
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 150-250 of human HCRTR1 (NP_001516.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hypocretin (orexin) receptor 1; hypocretin (orexin) receptor 1 hypocretin receptor 1 Hypocretin receptor type 1 hypocretin receptor-1 orexin receptor 1 orexin receptor type 1 orexin receptor-1 ox-1-R Ox1-R OX1R; Hypocretin receptor type 1; hypocretin receptor-1; orexin receptor 1; Orexin receptor type 1; orexin receptor-1; Ox-1-R; ox1-R
Gene Aliases: HCRTR1; OX1R
UniProt ID: (Human) O43613
Entrez Gene ID: (Human) 3061
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.