Novus Biologicals
Manufacturer Code:NBP211834AF488
Catalog # NB211834488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0501-5G21 |
Immunogen | Human Orexin B |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Hcrt; HCRT Hcrt 2 Hcrt2 Hypocretin Hypocretin (orexin) neuropeptide precursor Hypocretin 2 Hypocretin B Hypocretin precursor Hypocretin2 Orexin Orexin neuropeptide precursor Orexin precursor OrexinB PPORX PPOX; Hcrt1; Hcrt2; Hypocretin; hypocretin (orexin) neuropeptide; Hypocretin-1; Hypocretin-2; Orexin; Orexin-A; Orexin-B; prepro-orexin
Gene Aliases: HCRT; NRCLP1; OX; PPORX; PPOX
UniProt ID: (Human) O43612
Entrez Gene ID: (Human) 3060
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.