Invitrogen
Manufacturer Code:PA578389
Catalog # PIPA578389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human Orexin B. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hcrt; Hcrt1; Hcrt2; Hypocretin; Hypocretin (orexin) neuropeptide; Hypocretin-1; Hypocretin-2; Orexin; orexin A; orexin B; Orexin-A; Orexin-B; PPOX HCRT OX hypocretin (orexin) neuropeptide precursor NRCLP1; prepro-orexin
Gene Aliases: HCRT; NRCLP1; orexin-A; OX; PPORX; PPOX
UniProt ID: (Human) O43612, (Mouse) O55241, (Rat) O55232
Entrez Gene ID: (Human) 3060, (Mouse) 15171, (Rat) 25723
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.