R&D Systems
Manufacturer Code:MAB763100
Catalog # MAB763100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 97505 |
Immunogen | KLH-coupled human Orexin A/Hypocretin-1 synthetic peptide Accession # O43612 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Hcrt; HCRT Hcrt1 hypocretin (orexin) neuropeptide precursor Hypocretin1 Hypocretin-1 NRCLP1 orexin OXHcrt PPORX PPOXHypocretin; Hcrt1; Hcrt2; Hypocretin; hypocretin (orexin) neuropeptide; Hypocretin-1; Hypocretin-2; Orexin; Orexin-A; Orexin-B; prepro-orexin
Gene Aliases: HCRT; NRCLP1; OX; PPORX; PPOX
UniProt ID: (Human) O43612
Entrez Gene ID: (Human) 3060
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.