Invitrogen
Manufacturer Code:PA570436
Catalog # PIPA570436
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of rat Orexin A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hcrt; Hcrt1; Hcrt2; Hypocretin; Hypocretin (orexin) neuropeptide; Hypocretin-1; Hypocretin-1 Hcrt1; Hypocretin-2; Orexin; orexin A; orexin B; Orexin-A; Orexin-B; prepro-orexin
Gene Aliases: HCRT; NRCLP1; orexin-A; ORX; OX; PPORX; PPOX
UniProt ID: (Human) O43612, (Dog) Q9GLF6, (Pig) O77668, (Mouse) O55241, (Rat) O55232
Entrez Gene ID: (Horse) 100066236, (Rabbit) 100343676, (Bovine) 281222, (Human) 3060, (Dog) 607641, (Pig) 397305, (Sheep) 101122291, (Mouse) 15171, (Rat) 25723, (Guinea pig) 100717357
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.