Invitrogen
Manufacturer Code:PA118310
Catalog # 01-670-563
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues C(14) R L Y E L L H G A G N H A A G I L T L(33) of Bovin Orexin A conjugated to KLH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hcrt; Hcrt1; Hcrt2; Hypocretin; Hypocretin (orexin) neuropeptide; Hypocretin-1; Hypocretin-1 Hcrt1; Hypocretin-2; Orexin; Orexin-A; Orexin-B
Gene Aliases: Hcrt; orexin-A; Ox; Ppox
UniProt ID: (Rat) O55232
Entrez Gene ID: (Rat) 25723
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.