Invitrogen
Manufacturer Code:PA5107203
Catalog # PIPA5107203
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide located within the following region: CYLQVWLAIR AVAKQQKESE STQKAEKEVT RMVVVMVFAY CLCWGPYTFF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Green cone photoreceptor pigment; green long wavelength sensitive cone opsin; green LWS photopigment; Green-sensitive opsin; M opsin; Medium wavelength-sensitive cone opsin; Medium-wave-sensitive opsin 1; midwavelength sensitive opsin; MWS opsin
Gene Aliases: Gcp; Opn1mw
UniProt ID: (Mouse) O35599
Entrez Gene ID: (Mouse) 14539
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.