Invitrogen
Manufacturer Code:PA534041
Catalog # PIPA534041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 16 amino acid peptide from C-terminus of human OPN5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G-protein coupled receptor 136; G-protein coupled receptor PGR12; Neuropsin; OPN5 G-protein coupled receptor 136 Neuropsin Opsin 5 Opsin-5 PGR12 TMEM13 Transmembrane protein 13 DJ402H5.1 GPR136; Opsin-5; Transmembrane protein 13
Gene Aliases: GPR136; GRP136; OPN5; PGR12; TMEM13
UniProt ID: (Human) Q6U736
Entrez Gene ID: (Human) 221391
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.