Invitrogen
Manufacturer Code:OSR00218W
Catalog # PIOSR00218W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from mouse opsin-5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 136; G-protein coupled receptor 136; G-protein coupled receptor PGR12; Neuropsin; Opsin-5; Opsin-5 neuropsin G-protein coupled receptor 136 G-protein coupled receptor PGR12 OPN5 opsin 5 GPR136 TMEM13; Transmembrane protein 13
Gene Aliases: GPR136; GRP136; Neuropsin; OPN5; PGR12; TMEM13
UniProt ID: (Human) Q6U736, (Mouse) Q6VZZ7
Entrez Gene ID: (Human) 221391, (Rat) 316259, (Mouse) 353344
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.