Novus Biologicals
Manufacturer Code:NBP16903820UL
Catalog # NBP16903820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to OPN5 (opsin 5) The peptide sequence was selected form the C terminal of OPN5. Peptide sequence FFCLLLPTAVIVFSYVKIIAKVKSSSKEVAHFDSRIHSSHVLEMKLTKVA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dJ402H5.1 GPR136 G-protein coupled receptor 136 G-protein coupled receptor PGR12 Neuropsin opsin 5 opsin-5 PGR12 TMEM13 Transmembrane protein 13neuropsin; G-protein coupled receptor 136; G-protein coupled receptor PGR12; Neuropsin; Opsin-5; Transmembrane protein 13
Gene Aliases: GPR136; GRP136; OPN5; PGR12; TMEM13
UniProt ID: (Human) Q6U736
Entrez Gene ID: (Human) 221391
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.