Novus Biologicals
Manufacturer Code:NBP2298580.025MG
Catalog # NB019488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides from mouse green opsin and zebra finch red opsin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: cone dystrophy 5 (X-linked); GCP GOP Green cone photoreceptor pigment Green-sensitive opsin medium-wave-sensitive opsin 1 OPN1MW opsin 1 (cone pigments) medium-wave-sensitive 2; GOP; Green cone photoreceptor pigment; green cone pigment; Green-sensitive opsin; Medium-wave-sensitive opsin 1; Medium-wave-sensitive opsin 2; Medium-wave-sensitive opsin 3; Opsin 1 cone pigments medium-wave-sensitive 2; opsin 1 cone pigments medium-wave-sensitive 3; photopigment apoprotein
Gene Aliases: CBBM; CBD; COD5; GCP; GOP; OPN1MW; OPN1MW1; OPN1MW2; OPN1MW3
UniProt ID: (Human) P0DN78
Entrez Gene ID: (Human) 2652
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.