Proteintech
Manufacturer Code:110761AP150UL
Catalog # 50-172-6854
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Oligophrenin 1 Fusion Protein Ag1536 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mental retardation, X-linked 60; MRX60 oligophrenin 1 OPHN1 OPN1; Oligophrenin-1; oligophrenin-1, Rho-GTPase activating protein
Gene Aliases: ARHGAP41; C130037N19Rik; MRX60; OPHN1; OPN1
UniProt ID: (Human) O60890, (Mouse) Q99J31
Entrez Gene ID: (Human) 4983, (Mouse) 94190
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.