R&D Systems
Manufacturer Code:BAF2356
Catalog # BAF2356
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Olig3 Met1-Lys272 Accession # Q7RTU3 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHb7; Bhlhb7 BHLHE20 bHLHe20oligo3 Class B basic helix-loop-helix protein 7 Class E basic helix-loop-helix protein 20 Oligo3 oligodendrocyte transcription factor 3; bHLHe20; Class B basic helix-loop-helix protein 7; Class E basic helix-loop-helix protein 20; Oligo3; Oligodendrocyte transcription factor 3
Gene Aliases: BHLHB7; BHLHE20; OLIG3
UniProt ID: (Human) Q7RTU3
Entrez Gene ID: (Human) 167826
Molecular Function:
basic helix-loop-helix transcription factor
nuclease
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.