Novus Biologicals
Manufacturer Code:NBP128667
Catalog # NBP128667
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant mouse Olig2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic domain, helix-loop-helix protein, class B, 1; bHLHb1; BHLHB1 BHLHE19 Class B basic helix-loop-helix protein 1 Class E basic helix-loop-helix protein 19 human protein kinase C-binding protein RACK17 OLIGO2 oligodendrocyte lineage transcription factor 2 oligodendrocyte transcription factor 2 oligodendrocyte-specific bHLH transcription factor 2 protein kinase C binding protein 2 Protein kinase C-binding protein 2 Protein kinase C-binding protein RACK17 RACK17helix-loop-helix protein class B 1; bHLHe19; Class B basic helix-loop-helix protein 1; Class E basic helix-loop-helix protein 19; human protein kinase C-binding protein RACK17; Oligo2; Oligodendrocyte transcription factor 2; oligodendrocyte-specific bHLH transcription factor 2; Protein kinase C-binding protein 2; Protein kinase C-binding protein RACK17
Gene Aliases: BHLHB1; BHLHE19; OLIG2; OLIGO2; PRKCBP2; RACK17
UniProt ID: (Human) Q13516
Entrez Gene ID: (Human) 10215
Molecular Function:
basic helix-loop-helix transcription factor
nuclease
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.