Invitrogen
Manufacturer Code:OSR00035G
Catalog # PIOSR00035G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from mouse olfactory receptor family 42 subfamily 3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G-protein-coupled receptor; GA_x6K02T2PBJ9-5206624-5205620; odorant receptor S6; Odorant receptor S6 Olfactory receptor 544 Olfactory receptor Olfr544 ORL471 S79 ORL463. GENE NAME: Olfr544.; odorant receptor S79; olfactory receptor 544; olfactory receptor MOR42-3
Gene Aliases: MOR42-3; Ors6
Entrez Gene ID: (Mouse) 257926
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.